Assay Details
| Gene Symbol: Dclk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195496.1 | NP_001182425.1 | 7 - 8 | 1883 | 62 | |
| NM_001195497.1 | NP_001182426.1 | 6 - 7 | 1835 | 62 | ||
| NM_001195498.1 | NP_001182427.1 | 7 - 8 | 1886 | 62 | ||
| NM_001195499.1 | NP_001182428.1 | 7 - 8 | 1883 | 62 | ||
| NM_001195500.1 | NP_001182429.1 | 6 - 7 | 1835 | 62 | ||
| NM_027539.5 | NP_081815.3 | 6 - 7 | 1838 | 62 | ||
| GenBank mRNA | AB198721.1 | - | 7 - 8 | 1240 | 62 | |
| AK049126.1 | - | 6 - 7 | 1325 | 62 | ||
| AK049179.1 | - | 6 - 7 | 1287 | 62 | ||
| AK082633.1 | - | 6 - 7 | 1334 | 62 | ||
| AY968049.1 | - | 7 - 8 | 1171 | 62 | ||
| AY968050.1 | - | 7 - 8 | 1174 | 62 | ||
| AY968051.1 | - | 7 - 8 | 1171 | 62 | ||
| BC056921.1 | - | 6 - 7 | 1806 | 6822681 | 62 | |
| BC060192.1 | - | 6 - 7 | 1139 | 62 | ||
More Information
Set Membership: |
|

Dclk2
Genomic Map