Assay Details
| Gene Symbol: Prdm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027547.2 | NP_081823.2 | 12 - 13 | 1581 | 66 | |
| GenBank mRNA | AK014501.1 | - | 12 - 13 | 1505 | 66 | |
| AK018310.1 | - | 8 - 9 | 967 | 66 | ||
| AK021384.1 | - | 3 - 4 | 390 | 66 | ||
| BC138901.1 | - | 12 - 13 | 1555 | 8861923 | 66 | |
| BC138902.1 | - | 12 - 13 | 1555 | 8861924 | 66 | |
| BG070775.2 | - | 3 - 4 | 66 | |||
| BX511446.1 | - | 1 - 2 | 84 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prdm5
Genomic Map