Assay Details
| Gene Symbol: Sept12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027669.3 | NP_081945.1 | 6 - 7 | 761 | 69 | |
| NM_029374.1 | NP_083650.1 | 6 - 7 | 761 | 69 | ||
| GenBank mRNA | AK006458.1 | - | 6 - 7 | 699 | 69 | |
| AK016798.1 | - | 6 - 7 | 689 | 69 | ||
| AK077091.1 | - | 6 - 7 | 762 | 69 | ||
| BC115618.1 | - | 6 - 7 | 672 | 40102763 | 69 | |
| BC115619.1 | - | 5 - 6 | 807 | 40102765 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sept12
Genomic Map