Assay Details
| Gene Symbol: Daw1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027725.3 | NP_082001.2 | 7 - 8 | 691 | 116 | |
| NR_027636.1 | 10 - 11 | 986 | 116 | |||
| GenBank mRNA | AK016201.1 | - | 10 - 11 | 981 | 116 | |
| AK016977.1 | - | 7 - 8 | 692 | 116 | ||
| BC049704.1 | - | 7 - 8 | 691 | 116 | ||
| BC144994.1 | - | 7 - 8 | 668 | 116 | ||
| BC145995.1 | - | 7 - 8 | 691 | 40131298 | 116 | |
More Information
Set Membership: |
|

Daw1
Genomic Map