| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1000001122.mm.1 |
NM_027930.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000001122.mm.1 |
NM_027930.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000001122.mm.1 |
TC1000000158.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000001123.mm.1 |
NM_027930.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000001123.mm.1 |
NM_027930.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000001123.mm.1 |
TC1000000158.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000001124.mm.1 |
NM_027930.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000001124.mm.1 |
NM_027930.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000001124.mm.1 |
TC1000000158.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000001120.mm.1 |
NM_027930.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000001120.mm.1 |
NM_027930.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000001120.mm.1 |
TC1000000158.mm.1 |
MTA TC |