Assay Details
| Gene Symbol: Mtfr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027930.3 | NP_082206.2 | 5 - 6 | 657 | 87 | |
| GenBank mRNA | AK011410.1 | - | 5 - 6 | 652 | 87 | |
| AK016786.1 | - | 5 - 6 | 702 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26986426 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26986428 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26986425 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26986431 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26986427 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26986429 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1997300 | NM_027930.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P109460 | NM_027930.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P109460 | AK011410.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P109460 | AK016786.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 962773 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 87505 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 545693 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 387370 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 211906 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 638091 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 526886 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 548735 | AJ430350.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 548735 | AK076434.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 548735 | NM_173379.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 136718 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P154931 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P119401 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P109460 | NM_027930.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P109460 | AK011410.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P109460 | AK016786.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1997300 | NM_027930.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P116289 | BC019132.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P116289 | NM_027930.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P116289 | AK011410.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26986439 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26986398 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26986424 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26986431 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26986412 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26986432 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26986436 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26986422 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26986405 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26986433 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26986413 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26986428 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26986402 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26986419 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26986438 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26986406 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26986417 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26986425 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26986415 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26986409 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26986400 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26986403 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26986407 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26986399 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26986434 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06995089 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P154931 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P119401 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06995099 | - | - |
| Whole Mouse Genome G4122A | A_51_P116289 | BC019132.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P116289 | NM_027930.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P116289 | AK011410.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P681495 | BC019132.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P681495 | NM_027930.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P681495 | AK011410.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P681495 | AK016786.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121138 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109460 | NM_027930.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109460 | AK011410.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109460 | AK016786.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P154931 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P119401 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08957950 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08957947 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08957949 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08957948 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26986425 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26986427 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26986429 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26986428 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26986426 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26986431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26986518 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26986412 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26986406 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08957946 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26986420 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26986409 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26986463 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26986465 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08378448 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26986434 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26986425 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26986432 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26986449 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26986452 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08322950 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08378449 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26986402 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26986440 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26986455 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26986437 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26986447 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26986421 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26986441 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26986446 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08957947 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26986462 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26986407 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26986464 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08253818 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26986400 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08601961 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26986431 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26986460 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26986415 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26986417 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26986450 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08343641 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26986436 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08253822 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08335078 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26986445 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26986442 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26986404 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26986457 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26986439 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26986438 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10361997 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10361999 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10362002 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10362001 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10361998 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10361996 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10362000 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1432013_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10361995 | - | - |
| Mouse Genome 430 2.0 Array | 1432013_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1432013_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71804_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71804_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000001125.mm.1 | NM_027930.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000001125.mm.1 | NM_027930.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000001125.mm.1 | TC1000000158.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000001130.mm.1 | NM_027930.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000001130.mm.1 | NM_027930.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000001130.mm.1 | TC1000000158.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000001126.mm.1 | NM_027930.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000001126.mm.1 | NM_027930.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000001126.mm.1 | TC1000000158.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000001129.mm.1 | NM_027930.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000001129.mm.1 | NM_027930.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000001129.mm.1 | TC1000000158.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000001127.mm.1 | NM_027930.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000001127.mm.1 | NM_027930.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000001127.mm.1 | TC1000000158.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1432013_a_at | - | - |
| Mouse Expression Set 430 | 1432013_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1432013_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1432013_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1432013_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1432013_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 452235 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4570195 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4570195 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004570195 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1237496 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1237496 | - | - |
| mouse-6 v1.1 (Illumina) | scl39100.8.1_73-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4570195 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl39100.8.1_73-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4570195 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2758602 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2758602 | - | - |

Mtfr2
Genomic Map