Assay Details
| Gene Symbol: Dpep3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027960.2 | NP_082236.1 | 3 - 4 | 664 | 79 | |
| GenBank mRNA | AF488553.1 | - | 3 - 4 | 545 | 79 | |
| AK006085.1 | - | 3 - 4 | 665 | 79 | ||
| BC051148.1 | - | 3 - 4 | 674 | 6704315 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dpep3
Genomic Map