Assay Details
| Gene Symbol: Dpep3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027960.2 | NP_082236.1 | 6 - 7 | 1050 | 126 | |
| GenBank mRNA | AF488553.1 | - | 6 - 7 | 931 | 126 | |
| AK006085.1 | - | 6 - 7 | 1051 | 126 | ||
| BC051148.1 | - | 6 - 7 | 1060 | 6704315 | 126 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dpep3
Genomic Map