Assay Details
| Gene Symbol: Dpep3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027960.2 | NP_082236.1 | 9 - 10 | 1342 | 61 | |
| GenBank mRNA | AF488553.1 | - | 9 - 10 | 1223 | 61 | |
| AK006085.1 | - | 9 - 10 | 1343 | 61 | ||
| BC051148.1 | - | 9 - 10 | 1352 | 6704315 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26137132 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26137126 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26137129 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26137128 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26137127 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26137131 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P329818 | AK006085.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P329818 | BC051148.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P329818 | NM_027960.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P329818 | AF488553.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P329818 | AK006085.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P329818 | BC051148.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P329818 | NM_027960.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P329818 | AF488553.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 488711 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 104705 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 440523 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 822826 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 117335 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P118791 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P329818 | AK006085.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P329818 | BC051148.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P329818 | NM_027960.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P329818 | AF488553.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P329818 | AK006085.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P329818 | BC051148.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P329818 | NM_027960.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P329818 | AF488553.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P329818 | AK006085.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P329818 | BC051148.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P329818 | NM_027960.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P329818 | AF488553.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P329818 | AK006085.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P329818 | BC051148.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P329818 | NM_027960.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P329818 | AF488553.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26137113 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26137144 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26137111 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26137133 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26137115 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26137128 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26137120 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26137135 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26137140 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26137138 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26137149 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26137136 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26137129 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26137141 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26137117 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26137122 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26137112 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26137126 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26137114 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26137131 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26137132 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26137124 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P118791 | - | - |
| Whole Mouse Genome G4122A | A_51_P329818 | AK006085.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P329818 | BC051148.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P329818 | NM_027960.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P329818 | AF488553.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P329818 | AK006085.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P329818 | BC051148.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P329818 | NM_027960.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P329818 | AF488553.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108135 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P118791 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06818401 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06719750 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06967747 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06719740 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06719739 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06719744 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26137132 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26137131 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26137127 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26137126 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26137128 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26137129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26137136 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137105 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137138 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06967747 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137101 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07069492 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137141 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137146 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137103 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137099 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06755988 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137096 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137124 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137115 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137110 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137117 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07431488 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137106 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07190638 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137133 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137135 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07019874 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137111 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137128 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137095 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137097 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06950317 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137114 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137107 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137144 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137126 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137131 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137122 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06719763 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137149 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26137120 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10581427 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581430 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581423 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581426 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581428 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581422 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581425 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581433 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581429 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581431 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581424 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581432 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429035_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10581421 | - | - |
| Mouse Genome 430 2.0 Array | 1429035_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1429035_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71854_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71854_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800021935.mm.1 | NM_027960.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800021935.mm.1 | NM_027960.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800021935.mm.1 | TC0800002846.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1429035_PM_at | - | - |
| Mouse Expression Set 430 | 1429035_at | - | - |
| Murine Genome U74v2 Set | 161474_r_at | - | - |
| Murine Genome U74v2 Set | 161474_PM_r_at | - | - |
| Murine Genome U74v2 Set | 161358_r_at | - | - |
| Murine Genome U74v2 Set | 161358_PM_r_at | - | - |
| Murine Genome U74v2 Set | 170160_PM_r_at | - | - |
| Murine Genome U74v2 Set | 170160_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1429035_at | - | - |
| Mouse Genome 430A 2.0 Array | 1429035_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1429035_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1429035_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 352654 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3710706 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3710706 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003710706 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2685120 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2685120 | - | - |
| mouse-6 v1.1 (Illumina) | scl34382.10.1_100-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3710706 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl34382.10.1_100-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3710706 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2685120 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2685120 | - | - |

Dpep3
Genomic Map