Assay Details
| Gene Symbol: Cers5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028015.2 | NP_082291.1 | 2 - 3 | 443 | 84 | |
| GenBank mRNA | AK010241.1 | - | 2 - 3 | 371 | 84 | |
| AK075694.1 | - | 2 - 3 | 363 | 84 | ||
| AK145589.1 | - | 2 - 3 | 351 | 84 | ||
| AK145762.1 | - | 2 - 3 | 365 | 84 | ||
| AK146220.1 | - | 2 - 3 | 365 | 84 | ||
| AK146417.1 | - | 2 - 3 | 334 | 84 | ||
| AK219835.1 | - | 2 - 3 | 153 | 84 | ||
| AY029533.1 | - | 2 - 3 | 363 | 84 | ||
| BC043059.1 | - | 390 | 6419704 | 84 | ||
| BC046797.1 | - | 2 - 3 | 443 | 6403129 | 84 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK146220.1 transcripts.
Set Membership: |
|

Cers5
Genomic Map