Assay Details
| Gene Symbol: Smg9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028047.2 | NP_082323.1 | 9 - 10 | 1260 | 67 | |
| GenBank mRNA | AK005126.1 | - | 9 - 10 | 1260 | 67 | |
| AK039368.1 | - | 9 - 10 | 1259 | 67 | ||
| AK141576.1 | - | 9 - 10 | 1251 | 67 | ||
| AK169651.1 | - | 9 - 10 | 1184 | 67 | ||
| BC034211.1 | - | 9 - 10 | 1175 | 4988361 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Smg9
Genomic Map