Assay Details
| Gene Symbol: Cnpy3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF361644.1 | - | 2 - 3 | 366 | 68 | |
| AK005532.1 | - | 2 - 3 | 349 | 68 | ||
| AK031742.1 | - | 2 - 3 | 204 | 68 | ||
| AK049358.1 | - | 2 - 3 | 352 | 68 | ||
| AK085617.1 | - | 2 - 3 | 359 | 68 | ||
| AK086940.1 | - | 2 - 3 | 299 | 68 | ||
| AK143428.1 | - | 2 - 3 | 342 | 68 | ||
| AK220209.1 | - | 2 - 3 | 176 | 68 | ||
| BB857917.1 | - | 2 - 3 | 369 | 68 | ||
| BC013549.1 | - | 2 - 3 | 368 | 4222133 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cnpy3
Genomic Map