Assay Details
| Gene Symbol: Cnpy3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF361644.1 | - | 3 - 4 | 457 | 62 | |
| AK005532.1 | - | 3 - 4 | 440 | 62 | ||
| AK031742.1 | - | 3 - 4 | 295 | 62 | ||
| AK049358.1 | - | 3 - 4 | 443 | 62 | ||
| AK082749.1 | - | 1 - 2 | 59 | 62 | ||
| AK085617.1 | - | 3 - 4 | 450 | 62 | ||
| AK086940.1 | - | 3 - 4 | 390 | 62 | ||
| AK143428.1 | - | 3 - 4 | 433 | 62 | ||
| AK220209.1 | - | 3 - 4 | 267 | 62 | ||
| BC013549.1 | - | 3 - 4 | 459 | 4222133 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cnpy3
Genomic Map