Assay Details
| Gene Symbol: Osgepl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285839.1 | NP_001272768.1 | 2 - 3 | 485 | 65 | |
| NR_104361.1 | 2 - 3 | 485 | 65 | |||
| GenBank mRNA | AK011265.1 | - | 2 - 3 | 433 | 65 | |
| AK045669.1 | - | 2 - 3 | 447 | 65 | ||
| AK137332.1 | - | 2 - 3 | 436 | 65 | ||
| BC058172.1 | - | 2 - 3 | 448 | 5012054 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P121 | BC051211.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P121 | BC030671.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P121 | BC038910.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P121 | AK045669.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P121 | AK137332.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P121 | BC051211.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P121 | BC030671.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P121 | BC038910.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P121 | AK045669.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P121 | AK137332.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 184476 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 618449 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 224302 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 18223 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 367960 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 572737 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 460091 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 48679 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P180185 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P149585 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P121 | BC051211.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P121 | BC030671.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P121 | BC038910.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P121 | AK045669.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P121 | AK137332.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P121 | BC051211.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P121 | BC030671.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P121 | BC038910.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P121 | AK045669.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P121 | AK137332.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20231311 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20231344 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20231335 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20231342 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20231333 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20231332 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20231339 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20231330 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20231313 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20231321 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20231325 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20231315 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20231319 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20231331 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20231327 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20231340 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20231312 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20231323 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20231308 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20231314 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20231317 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20231328 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20231337 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P149585 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04098690 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P180185 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00068871 | - | - |
| Whole Mouse Genome G4122A | A_52_P121 | BC051211.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P121 | BC030671.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P121 | BC038910.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P121 | AK045669.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P121 | AK137332.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P121 | BC051211.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P121 | BC030671.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P121 | BC038910.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P121 | AK045669.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P121 | AK137332.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119894 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P149585 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P180185 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00227410 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00227414 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00227393 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00227400 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00227415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20231376 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231454 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231388 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231445 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231447 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231344 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231384 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231304 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231376 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00079415 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00330785 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231409 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231402 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231317 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231312 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231437 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231346 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231416 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231397 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231340 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00365456 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231440 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231337 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231349 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231396 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231355 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231350 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231351 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231410 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231368 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231385 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231370 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231405 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231381 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00227400 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231308 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00835396 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231452 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231364 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00674264 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231377 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231333 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231313 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231451 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231392 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231331 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231357 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00227410 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231358 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231375 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231367 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231330 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231378 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231362 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231400 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231401 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00021420 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231407 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231323 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231366 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231342 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00021419 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00265923 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231403 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231319 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231393 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231321 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231394 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231332 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231379 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00227394 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231314 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231360 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00227413 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20231386 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00227412 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10346266 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346264 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346268 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346263 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346265 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346267 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346262 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10346261 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1435127_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428558_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439715_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428557_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10346260 | - | - |
| Mouse Genome 430 2.0 Array | 1439715_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1435127_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1435127_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1428557_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1428558_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1428557_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1439715_at | - | - |
| Mouse Genome 430 2.0 Array | 1428558_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72085_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72085_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100003017.mm.1 | NM_001285839.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100003017.mm.1 | NR_104360.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100003017.mm.1 | NR_104361.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100003017.mm.1 | TC0100000387.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1428557_a_at | - | - |
| Mouse Expression Set 430 | 1439715_PM_at | - | - |
| Mouse Expression Set 430 | 1428557_PM_a_at | - | - |
| Mouse Expression Set 430 | 1435127_PM_a_at | - | - |
| Mouse Expression Set 430 | 1428558_PM_at | - | - |
| Mouse Expression Set 430 | 1435127_a_at | - | - |
| Mouse Expression Set 430 | 1439715_at | - | - |
| Mouse Expression Set 430 | 1428558_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439715_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428557_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428558_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435127_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435127_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428558_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428557_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439715_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 671485 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 460731 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 460731 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1340014 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000460731 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001340014 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1226615 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1225723 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1226615 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1225723 | - | - |
| mouse-6 v1.1 (Illumina) | scl0072085.2_165-S | - | - |
| mouse-6 v1.1 (Illumina) | scl072085.4_41-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 460731 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1340014 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0072085.2_165-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl072085.4_41-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 460731 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1340014 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1225723 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1226615 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1225723 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1226615 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl072085.4_41-S | - | - |

Osgepl1
Genomic Map