Assay Details
| Gene Symbol: Osgepl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285839.1 | NP_001272768.1 | 5 - 6 | 1353 | 68 | |
| GenBank mRNA | AK011265.1 | - | 5 - 6 | 1301 | 68 | |
| AK045669.1 | - | 5 - 6 | 1315 | 68 | ||
| BC038910.1 | - | 3 - 4 | 679 | 68 | ||
| BC051211.1 | - | 2 - 3 | 729 | 68 | ||
| BC058172.1 | - | 5 - 6 | 1316 | 5012054 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Osgepl1
Genomic Map