Assay Details
| Gene Symbol: Mfsd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028140.4 | NP_082416.2 | 5 - 6 | 705 | 82 | |
| GenBank mRNA | AK013144.1 | - | 5 - 6 | 597 | 82 | |
| AK032080.1 | - | 5 - 6 | 726 | 82 | ||
| AK035291.1 | - | 5 - 6 | 668 | 82 | ||
| AK077957.1 | - | 5 - 6 | 706 | 82 | ||
| BC010483.1 | - | 5 - 6 | 554 | 82 | ||
| BC113183.1 | - | 5 - 6 | 636 | 40061891 | 82 | |
| BC113790.1 | - | 5 - 6 | 636 | 40061889 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P323148 | BC010483.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P323148 | AK035291.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P323148 | AK077957.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P323148 | NM_028140.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P323148 | BC010483.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P323148 | AK035291.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P323148 | AK077957.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P323148 | NM_028140.4 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 27058 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 699360 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 66588 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 752863 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 477609 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 370118 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 471448 | AK012533.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 471448 | AK012762.2 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 471448 | AK014084.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 471448 | AK163310.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 471448 | BC079900.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 471448 | NM_025642.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 213987 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 190633 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 489559 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 936661 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 136860 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 919666 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 647975 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 127406 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 142215 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 91396 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P152999 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P133416 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P323148 | BC010483.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P323148 | AK035291.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P323148 | AK077957.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P323148 | NM_028140.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P323148 | BC010483.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P323148 | AK035291.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P323148 | AK077957.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P323148 | NM_028140.4 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22081851 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22081824 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22081834 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P133416 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04883785 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P152999 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00625583 | - | - |
| Whole Mouse Genome G4122A | A_52_P660405 | BC113183.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P660405 | BC113790.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P660405 | BC010483.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P660405 | AK035291.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P660405 | AK077957.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P623352 | AK084485.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P323148 | BC010483.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P323148 | AK035291.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P323148 | AK077957.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P323148 | NM_028140.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P323148 | BC010483.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P323148 | AK035291.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P323148 | AK077957.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P323148 | NM_028140.4 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128126 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122977 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P133416 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P152999 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02445035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22081785 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22081780 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22081861 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22081879 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22081811 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22081814 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02856081 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22081670 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22081828 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22081664 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22081841 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22081851 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22081777 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22081785 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22081824 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22081673 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22081784 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22081781 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22081671 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22081877 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22081775 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22081667 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22081812 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22081672 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22081675 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22081857 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22081669 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22081786 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10497956 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10497945 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10497954 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10497957 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10497947 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10497949 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10497955 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10497952 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10497953 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10497948 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10497951 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10497946 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10497950 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438497_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452335_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10497944 | - | - |
| Mouse Genome 430 2.0 Array | 1452335_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1438497_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1438497_at | - | - |
| Mouse Genome 430 2.0 Array | 1452335_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72175_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72175_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300014607.mm.1 | NM_028140.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300014607.mm.1 | NM_028140.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300014607.mm.1 | TC0300001902.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1452335_at | - | - |
| Mouse Expression Set 430 | 1452335_PM_at | - | - |
| Mouse Expression Set 430 | 1438497_PM_at | - | - |
| Mouse Expression Set 430 | 1438497_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452335_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452335_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452335_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452335_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438497_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438497_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 916938 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1190390 | - | - |
| MouseWG-6 v2 Expression BeadChip | 102230504 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1190390 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 102230504 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001190390 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2635968 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2635968 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1222624 | - | - |
| mouse-6 v1.1 (Illumina) | scl22218.13_11-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102230504 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1190390 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl22218.13_11-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102230504 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1190390 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2635968 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2635968 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2578888 | - | - |

Mfsd8
Genomic Map