Assay Details
| Gene Symbol: Tmem175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163531.1 | NP_001157003.1 | 4 - 5 | 477 | 109 | |
| NM_001163532.1 | NP_001157004.1 | 4 - 5 | 480 | 109 | ||
| NM_028223.3 | NP_082499.3 | 4 - 5 | 587 | 109 | ||
| GenBank mRNA | AI526594.1 | - | 4 - 5 | 451 | 109 | |
| AK013879.1 | - | 4 - 5 | 410 | 109 | ||
| AK034579.1 | - | 4 - 5 | 456 | 109 | ||
| AK038865.1 | - | 4 - 5 | 447 | 109 | ||
| AK049952.1 | - | 4 - 5 | 521 | 109 | ||
| AK082765.1 | - | 4 - 5 | 443 | 109 | ||
| AK170273.1 | - | 4 - 5 | 430 | 109 | ||
| BB653267.1 | - | 4 - 5 | 517 | 109 | ||
| BC005542.1 | - | 4 - 5 | 395 | 3590497 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23973162 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1989573 | BC005542.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1989573 | AK082765.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1989573 | AK170273.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1989573 | AK034579.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1989573 | NM_028223.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2416862 | AK076446.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 652428 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 966394 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 19019 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 700197 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 316482 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 623859 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 695260 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 595833 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 641710 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 657638 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 88251 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 539957 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 927 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 94585 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P119069 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P135062 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1989573 | BC005542.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1989573 | AK082765.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1989573 | AK170273.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1989573 | AK034579.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1989573 | NM_028223.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2416862 | AK076446.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23973165 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23973176 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23973163 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23973166 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23973174 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23973169 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23973170 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23973172 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23973164 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23973171 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23973168 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23973177 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23973167 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05690258 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05690247 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P119069 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P135062 | - | - |
| Whole Mouse Genome G4122A | A_52_P548000 | AK038865.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P548000 | AK049952.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P456098 | AK132151.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P456098 | BC005542.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P456098 | AK034579.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P456098 | NM_028223.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P456098 | NM_001163531.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132821 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P135062 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P119069 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04600427 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04600454 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04600428 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23973162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23973230 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04600436 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973162 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973175 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973174 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973188 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973172 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973181 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973173 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973179 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04882131 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973185 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973177 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973161 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973170 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973163 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04765302 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973176 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973165 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04937433 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973184 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04600454 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10524027 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10524026 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10524032 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10524023 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10524031 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10524024 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10524029 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10524030 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10524033 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10524021 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10524028 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10524025 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452134_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10524020 | - | - |
| Mouse Genome 430 2.0 Array | 1435109_at | - | - |
| Mouse Genome 430 2.0 Array | 1452134_at | - | - |
| Mouse Genome 430 2.0 Array | 1452134_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1435109_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72392_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72392_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500009464.mm.1 | TC0500001110.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500009464.mm.1 | NM_001163531.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500009464.mm.1 | NM_001163531.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500009464.mm.1 | NM_001163532.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500009464.mm.1 | NM_001163532.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500009464.mm.1 | NM_028223.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500009464.mm.1 | NM_028223.3 | Refseq |
| Mouse Expression Set 430 | 1452134_at | - | - |
| Mouse Expression Set 430 | 1452134_PM_at | - | - |
| Mouse Expression Set 430 | 1435109_at | - | - |
| Mouse Expression Set 430 | 1435109_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452134_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452134_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452134_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452134_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 612453 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 102940278 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 101770019 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3850390 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003850390 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2677197 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2646438 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1215917 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2646438 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2677197 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1236468 | - | - |
| mouse-6 v1.1 (Illumina) | scl0072392.2_133-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102940278 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 101770019 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3850390 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0072392.2_133-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102940278 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3850390 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101770019 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2677197 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1236468 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1215917 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2677197 | - | - |

Tmem175
Genomic Map