Assay Details
| Gene Symbol: Ttc39c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028341.4 | NP_082617.2 | 5 - 6 | 1157 | 73 | |
| GenBank mRNA | AK013267.1 | - | 5 - 6 | 892 | 73 | |
| AK077971.1 | - | 217 | 73 | |||
| BC020021.1 | - | 5 - 6 | 907 | 2648772 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ttc39c
Genomic Map