Assay Details
| Gene Symbol: Ttc39c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028341.4 | NP_082617.2 | 9 - 10 | 1640 | 101 | |
| GenBank mRNA | AK013267.1 | - | 9 - 10 | 1375 | 101 | |
| AK077971.1 | - | 700 | 101 | |||
| BC020021.1 | - | 9 - 10 | 1390 | 2648772 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ttc39c
Genomic Map