Assay Details
| Gene Symbol: Ttc19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028360.2 | NP_082636.3 | 5 - 6 | 524 | 79 | |
| NM_029704.2 | NP_083980.2 | 4 - 5 | 485 | 79 | ||
| GenBank mRNA | AK013368.1 | - | 4 - 5 | 477 | 79 | |
| AK034075.1 | - | 5 - 6 | 514 | 79 | ||
| BC026866.1 | - | 3 - 4 | 168 | 79 | ||
| BC037111.1 | - | 4 - 5 | 248 | 5370464 | 79 | |
More Information
Set Membership: |
|

Ttc19
Genomic Map