Assay Details
| Gene Symbol: Calr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028500.3 | NP_082776.2 | 1 - 2 | 187 | 95 | |
| GenBank mRNA | AB206786.1 | - | 1 - 2 | 92 | 95 | |
| AK006582.1 | - | 1 - 2 | 188 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Calr3
Genomic Map