Assay Details
| Gene Symbol: Zbtb8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028603.4 | NP_082879.1 | 1 - 2 | 291 | 128 | |
| GenBank mRNA | AK010736.1 | - | 1 - 2 | 290 | 126 | |
| BC017614.1 | - | 1 - 2 | 122 | 4912757 | 128 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK010736.1,BC017614.1,NM_028603.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23296398 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23296404 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23296403 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23296400 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23296401 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23296399 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23296405 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P435817 | BC017614.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P435817 | NM_028603.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P435817 | AK010736.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P435817 | BC017614.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P435817 | NM_028603.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P435817 | AK010736.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 196893 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 830915 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 358977 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 937419 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 692741 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 13834 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 431596 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 154495 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 277941 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 958284 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 930711 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 774060 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 114121 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 182169 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 557490 | AK018320.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 67901 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 35968 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P132154 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P116687 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P435817 | BC017614.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P435817 | NM_028603.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P435817 | AK010736.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P435817 | BC017614.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P435817 | NM_028603.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P435817 | AK010736.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23296402 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23296408 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23296406 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23296396 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23296397 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23296409 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23296394 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23296410 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23296398 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23296413 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23296423 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23296416 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23296401 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23296415 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23296418 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23296421 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23296405 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23296419 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23296412 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00986723 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P116687 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P132154 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00986715 | - | - |
| Whole Mouse Genome G4122A | A_51_P435817 | BC017614.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P435817 | NM_028603.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P435817 | AK010736.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P435817 | BC017614.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P435817 | NM_028603.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P435817 | AK010736.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115354 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P116687 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P132154 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03687871 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03687872 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03687877 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03687878 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03687870 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23296399 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23296401 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23296398 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23296405 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23296404 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23296403 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23296400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23296336 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23296430 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23296424 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03727191 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23296382 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03726218 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23296394 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23296395 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23296304 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23296409 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03687871 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04005671 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23296299 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23296423 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03942348 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23296303 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23296379 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03706243 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23296391 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23296422 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23296308 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23296413 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23296377 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23296410 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23296405 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23296392 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23296383 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03942352 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23296416 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23296396 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23296415 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23296403 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03755608 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23296408 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23296301 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23296297 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23296389 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03942347 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23296380 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23296298 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03662220 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23296398 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23296397 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03252267 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04093674 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23296295 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03731934 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23296429 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10516591 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516593 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516594 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516592 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516595 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452063_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10516590 | - | - |
| Mouse Genome 430 2.0 Array | 1452063_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1452063_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73680_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73680_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400029289.mm.1 | TC0400003593.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400029289.mm.1 | NM_028603.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400029289.mm.1 | NM_028603.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400029288.mm.1 | TC0400003593.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400029288.mm.1 | NM_028603.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400029288.mm.1 | NM_028603.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400029286.mm.1 | TC0400003593.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400029286.mm.1 | NM_028603.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400029286.mm.1 | NM_028603.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400029287.mm.1 | TC0400003593.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400029287.mm.1 | NM_028603.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400029287.mm.1 | NM_028603.4 | Refseq |
| Mouse Expression Set 430 | 1452063_PM_at | - | - |
| Mouse Expression Set 430 | 1452063_at | - | - |
| Murine Genome U74v2 Set | 135180_PM_at | - | - |
| Murine Genome U74v2 Set | 135180_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452063_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452063_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452063_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452063_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 760220 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4280100 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004280100 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2753530 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1240747 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2753530 | - | - |
| mouse-6 v1.1 (Illumina) | scl23783.5.1_1-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4280100 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl23783.5.1_1-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4280100 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2854121 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2854117 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2854121 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2854117 | - | - |

Zbtb8a
Genomic Map