Assay Details
| Gene Symbol: Psd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289602.1 | NP_001276531.1 | 12 - 13 | 2239 | 65 | |
| NM_028707.4 | NP_082983.3 | 12 - 13 | 2245 | 65 | ||
| NR_110349.1 | 12 - 13 | 2165 | 65 | |||
| GenBank mRNA | AK018116.1 | - | 7 - 8 | 701 | 65 | |
| AK045529.1 | - | 12 - 13 | 1976 | 65 | ||
| AK053835.1 | - | 12 - 13 | 1893 | 65 | ||
| AK149027.1 | - | 12 - 13 | 1975 | 65 | ||
| BC056437.1 | - | 8 - 9 | 744 | 65 | ||
| BC062930.1 | - | 11 - 12 | 1826 | 6855293 | 65 | |
| BC065053.1 | - | 12 - 13 | 2013 | 5709231 | 65 | |
| BC065055.1 | - | 12 - 13 | 1996 | 5720949 | 65 | |
| BC066026.1 | - | 12 - 13 | 2245 | 5721455 | 65 | |
| BC066036.1 | - | 12 - 13 | 2002 | 6837266 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Psd2
Genomic Map