Assay Details
| Gene Symbol: Glyr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079814.1 | NP_001073282.1 | 7 - 8 | 747 | 90 | |
| NM_028720.2 | NP_082996.2 | 7 - 8 | 747 | 90 | ||
| GenBank mRNA | AK014456.1 | - | 7 - 8 | 687 | 90 | |
| AK040841.1 | - | 7 - 8 | 697 | 90 | ||
| AK145277.1 | - | 7 - 8 | 698 | 90 | ||
| AK150349.1 | - | 7 - 8 | 691 | 90 | ||
| AK151832.1 | - | 7 - 8 | 694 | 90 | ||
| AK152586.1 | - | 7 - 8 | 694 | 90 | ||
| AK152887.1 | - | 7 - 8 | 691 | 90 | ||
| AK159476.1 | - | 7 - 8 | 701 | 90 | ||
| AK188170.1 | - | 2 - 3 | 133 | 90 | ||
| AK204179.1 | - | 4 - 5 | 332 | 90 | ||
| BC006893.1 | - | 7 - 8 | 681 | 3599477 | 90 | |
| CF174076.1 | - | 2 - 3 | 85 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Glyr1
Genomic Map