Assay Details
| Gene Symbol: Glyr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079814.1 | NP_001073282.1 | 8 - 9 | 791 | 63 | |
| NM_028720.2 | NP_082996.2 | 8 - 9 | 791 | 63 | ||
| GenBank mRNA | AK014456.1 | - | 8 - 9 | 731 | 63 | |
| AK040841.1 | - | 8 - 9 | 741 | 63 | ||
| AK140812.1 | - | 6 - 7 | 2389 | 63 | ||
| AK145277.1 | - | 8 - 9 | 742 | 63 | ||
| AK150349.1 | - | 8 - 9 | 735 | 63 | ||
| AK151832.1 | - | 8 - 9 | 738 | 63 | ||
| AK152586.1 | - | 8 - 9 | 738 | 63 | ||
| AK152887.1 | - | 8 - 9 | 735 | 63 | ||
| AK159476.1 | - | 8 - 9 | 745 | 63 | ||
| BC006893.1 | - | 8 - 9 | 725 | 3599477 | 63 | |
| CF174076.1 | - | 3 - 4 | 129 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Glyr1
Genomic Map