Assay Details
| Gene Symbol: Glyr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079814.1 | NP_001073282.1 | 12 - 13 | 1184 | 81 | |
| NM_028720.2 | NP_082996.2 | 12 - 13 | 1166 | 81 | ||
| GenBank mRNA | AK014456.1 | - | 12 - 13 | 1106 | 81 | |
| AK040841.1 | - | 12 - 13 | 1116 | 81 | ||
| AK145277.1 | - | 12 - 13 | 1117 | 81 | ||
| AK150349.1 | - | 12 - 13 | 1110 | 81 | ||
| AK152887.1 | - | 12 - 13 | 1110 | 81 | ||
| AK159476.1 | - | 12 - 13 | 1120 | 81 | ||
| BC006893.1 | - | 12 - 13 | 1100 | 3599477 | 81 | |
| CF174076.1 | - | 7 - 8 | 522 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Glyr1
Genomic Map