Assay Details
| Gene Symbol: Nphp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028721.3 | NP_082997.3 | 18 - 19 | 2797 | 84 | |
| GenBank mRNA | AK014393.1 | - | 3 - 4 | 357 | 84 | |
| AK133217.1 | - | 18 - 19 | 2798 | 84 | ||
| AK173327.1 | - | 18 - 19 | 2376 | 84 | ||
| AY259499.1 | - | 18 - 19 | 2734 | 84 | ||
| BC115724.1 | - | 18 - 19 | 2324 | 40086844 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nphp3
Genomic Map