Assay Details
| Gene Symbol: Ifltd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028742.2 | NP_083018.1 | 5 - 6 | 702 | 87 | |
| GenBank mRNA | AK016641.1 | - | 5 - 6 | 702 | 87 | |
| AK052206.1 | - | 3 - 4 | 781 | 87 | ||
| AY753326.1 | - | 3 - 4 | 537 | 87 | ||
| BC053420.1 | - | 4 - 5 | 664 | 6705208 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ifltd1
Genomic Map