| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1900001084.mm.1 |
NM_001164709.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001084.mm.1 |
NM_001164709.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001084.mm.1 |
NM_028769.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001084.mm.1 |
NM_028769.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001084.mm.1 |
TC1900000094.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900001081.mm.1 |
NM_028769.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001081.mm.1 |
NM_028769.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001081.mm.1 |
TC1900000094.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900001085.mm.1 |
NM_001164709.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001085.mm.1 |
NM_001164709.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001085.mm.1 |
NM_028769.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001085.mm.1 |
NM_028769.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001085.mm.1 |
TC1900000094.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900001083.mm.1 |
NM_001164709.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001083.mm.1 |
NM_001164709.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001083.mm.1 |
TC1900000094.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900001074.mm.1 |
NM_028769.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001074.mm.1 |
NM_028769.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001074.mm.1 |
TC1900000094.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900001075.mm.1 |
NM_028769.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001075.mm.1 |
NM_028769.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001075.mm.1 |
TC1900000094.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900001072.mm.1 |
NM_028769.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001072.mm.1 |
NM_028769.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001072.mm.1 |
TC1900000094.mm.1 |
MTA TC |