Assay Details
| Gene Symbol: Syvn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164709.1 | NP_001158181.1 | 12 - 13 | 1437 | 108 | |
| NM_028769.5 | NP_083045.4 | 12 - 13 | 1804 | 108 | ||
| GenBank mRNA | AK004688.2 | - | 12 - 13 | 1291 | 108 | |
| AK122558.1 | - | 9 - 10 | 2190 | 108 | ||
| AK143720.1 | - | 12 - 13 | 1318 | 109 | ||
| BC032931.1 | - | 1 - 2 | 90 | 108 | ||
| BC042199.1 | - | 12 - 13 | 1299 | 4953165 | 108 | |
| BC046829.1 | - | 12 - 13 | 1437 | 6811574 | 108 | |
| BC057917.1 | - | 12 - 13 | 1718 | 5136398 | 108 | |
| BC080722.1 | - | 12 - 13 | 1322 | 5323695 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Syvn1
Genomic Map