Assay Details
| Gene Symbol: Syvn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164709.1 | NP_001158181.1 | 13 - 14 | 1593 | 87 | |
| NM_028769.5 | NP_083045.4 | 13 - 14 | 1960 | 87 | ||
| GenBank mRNA | AK004688.2 | - | 13 - 14 | 1447 | 87 | |
| AK122558.1 | - | 10 - 11 | 2346 | 87 | ||
| AK143720.1 | - | 13 - 14 | 1474 | 87 | ||
| AK172097.1 | - | 8 - 9 | 2522 | 87 | ||
| BC032931.1 | - | 2 - 3 | 246 | 87 | ||
| BC042199.1 | - | 13 - 14 | 1455 | 4953165 | 87 | |
| BC046829.1 | - | 13 - 14 | 1593 | 6811574 | 87 | |
| BC057917.1 | - | 13 - 14 | 1874 | 5136398 | 87 | |
| BC080722.1 | - | 13 - 14 | 1478 | 5323695 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Syvn1
Genomic Map