Assay Details
| Gene Symbol: Slc35f5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028787.4 | NP_083063.2 | 4 - 5 | 641 | 74 | |
| GenBank mRNA | AK004892.2 | - | 4 - 5 | 622 | 74 | |
| AK145848.1 | - | 4 - 5 | 642 | 74 | ||
| AK169012.1 | - | 4 - 5 | 503 | 74 | ||
| AK171951.1 | - | 4 - 5 | 615 | 74 | ||
| BC026858.1 | - | 4 - 5 | 500 | 4014064 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc35f5
Genomic Map