Assay Details
| Gene Symbol: Stk40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145827.1 | NP_001139299.1 | 11 - 12 | 1560 | 60 | |
| NM_028800.3 | NP_083076.3 | 9 - 10 | 1206 | 60 | ||
| GenBank mRNA | AB102946.1 | - | 9 - 10 | 1037 | 60 | |
| AK009154.1 | - | 4 - 5 | 424 | 60 | ||
| AK050808.1 | - | 5 - 6 | 509 | 60 | ||
| AK154780.1 | - | 11 - 12 | 1551 | 60 | ||
| AY336057.1 | - | 9 - 10 | 1191 | 60 | ||
| BC046981.1 | - | 9 - 10 | 1165 | 6478265 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stk40
Genomic Map