Assay Details
| Gene Symbol: Gpcpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042672.1 | NP_001036137.1 | 14 - 15 | 1482 | 100 | |
| NM_001291050.1 | NP_001277979.1 | 13 - 14 | 1355 | 100 | ||
| NM_001291051.1 | NP_001277980.1 | 13 - 14 | 1385 | 100 | ||
| NM_001291052.1 | NP_001277981.1 | 13 - 14 | 1403 | 100 | ||
| NM_027096.2 | NP_081372.1 | 12 - 13 | 1279 | 100 | ||
| NM_028802.3 | NP_083078.3 | 14 - 15 | 1479 | 100 | ||
| GenBank mRNA | AK009563.1 | - | 7 - 8 | 698 | 100 | |
| AK030645.1 | - | 14 - 15 | 1433 | 100 | ||
| AK031658.1 | - | 13 - 14 | 1358 | 100 | ||
| AK049491.1 | - | 6 - 7 | 568 | 100 | ||
| AK050318.1 | - | 14 - 15 | 1394 | 100 | ||
| AK051728.1 | - | 6 - 7 | 542 | 100 | ||
| AK141374.1 | - | 14 - 15 | 1369 | 100 | ||
| AK163419.1 | - | 13 - 14 | 1326 | 100 | ||
| AK165952.1 | - | 14 - 15 | 1444 | 100 | ||
| AK166293.1 | - | 5 - 6 | 413 | 100 | ||
| AK166454.1 | - | 13 - 14 | 1368 | 100 | ||
| BC006887.1 | - | 13 - 14 | 1327 | 100 | ||
| BC033408.1 | - | 12 - 13 | 1209 | 5357683 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpcpd1
Genomic Map