Assay Details
| Gene Symbol: Tc2n | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082976.2 | NP_001076445.1 | 5 - 6 | 721 | 131 | |
| NM_001286364.1 | NP_001273293.1 | 5 - 6 | 1384 | 131 | ||
| NM_028924.3 | NP_083200.1 | 5 - 6 | 1073 | 131 | ||
| GenBank mRNA | AB062282.1 | - | 4 - 5 | 553 | 131 | |
| AK016683.1 | - | 5 - 6 | 1405 | 131 | ||
| AK136558.1 | - | 5 - 6 | 722 | 131 | ||
| BC120793.1 | - | 5 - 6 | 1073 | 40129716 | 131 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tc2n
Genomic Map