Assay Details
| Gene Symbol: Snx29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290148.1 | NP_001277077.1 | 11 - 12 | 1059 | 79 | |
| NM_028964.4 | NP_083240.2 | 18 - 19 | 2108 | 79 | ||
| GenBank mRNA | AF399755.1 | - | 11 - 12 | 1014 | 79 | |
| AK017102.1 | - | 11 - 12 | 1060 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Snx29
Genomic Map