Assay Details
| Gene Symbol: Sf3a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029157.3 | NP_083433.1 | 6 - 7 | 498 | 82 | |
| GenBank mRNA | AK030743.1 | - | 6 - 7 | 533 | 82 | |
| AK150733.1 | - | 6 - 7 | 496 | 82 | ||
| AK151141.1 | - | 6 - 7 | 496 | 82 | ||
| AK151382.1 | - | 6 - 7 | 496 | 82 | ||
| AK161646.1 | - | 6 - 7 | 519 | 82 | ||
| AK168966.1 | - | 6 - 7 | 522 | 82 | ||
| AK169324.1 | - | 6 - 7 | 542 | 82 | ||
| BC009141.1 | - | 6 - 7 | 498 | 3598647 | 82 | |
| BC092058.1 | - | 6 - 7 | 530 | 4017388 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sf3a3
Genomic Map