Assay Details
| Gene Symbol: Spag16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271533.1 | NP_001258462.1 | 4 - 5 | 410 | 138 | |
| NM_029160.3 | NP_083436.2 | 4 - 5 | 410 | 138 | ||
| GenBank mRNA | AF490391.1 | - | 4 - 5 | 392 | 138 | |
| AK015884.1 | - | 4 - 5 | 411 | 138 | ||
| AK083083.1 | - | 4 - 5 | 389 | 138 | ||
| AK162117.1 | - | 3 - 4 | 325 | 138 | ||
| BC120668.1 | - | 4 - 5 | 409 | 40129591 | 138 | |
| BC145006.1 | - | 4 - 5 | 409 | 9053554 | 138 | |
More Information
Updated Mapping Information
Set Membership: |
|

Spag16
Genomic Map