Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1800003885.mm.1 |
NM_029394.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800003885.mm.1 |
NM_029394.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800003885.mm.1 |
TC1800000535.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800003888.mm.1 |
NM_029394.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800003888.mm.1 |
NM_029394.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800003888.mm.1 |
TC1800000535.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800003886.mm.1 |
NM_029394.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800003886.mm.1 |
NM_029394.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800003886.mm.1 |
TC1800000535.mm.1 |
MTA TC |