Assay Details
| Gene Symbol: Tmem14a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290679.1 | NP_001277608.1 | 2 - 3 | 404 | 71 | |
| NM_029398.3 | NP_083674.2 | 2 - 3 | 404 | 71 | ||
| GenBank mRNA | AK017734.1 | - | 2 - 3 | 319 | 71 | |
| BC034768.1 | - | 2 - 3 | 350 | 4039224 | 71 | |
| BC060034.1 | - | 2 - 3 | 348 | 6491675 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem14a
Genomic Map