Assay Details
| Gene Symbol: Klf17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029416.2 | NP_083692.2 | 1 - 2 | 168 | 65 | |
| GenBank mRNA | AF508984.1 | - | 1 - 2 | 134 | 65 | |
| AK018361.1 | - | 1 - 2 | 134 | 65 | ||
| AK139827.1 | - | 1 - 2 | 135 | 65 | ||
| BC064748.1 | - | 1 - 2 | 112 | 30476790 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Klf17
Genomic Map