Assay Details
| Gene Symbol: Klf17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029416.2 | NP_083692.2 | 2 - 3 | 961 | 86 | |
| GenBank mRNA | AF508984.1 | - | 2 - 3 | 927 | 86 | |
| AK018361.1 | - | 2 - 3 | 927 | 86 | ||
| AK139827.1 | - | 2 - 3 | 928 | 86 | ||
| BC064748.1 | - | 2 - 3 | 905 | 30476790 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
gamete generation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
nucleic acid binding
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription regulatory region DNA binding
metal ion binding
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
gamete generation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
nucleic acid binding
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription regulatory region DNA binding
metal ion binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_65_P18756 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P271068 | BC064748.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P271068 | AK018361.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P271068 | AK139827.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P271068 | NM_029416.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P271068 | AF508984.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P271068 | BC064748.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P271068 | AK018361.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P271068 | AK139827.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P271068 | NM_029416.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P271068 | AF508984.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 286962 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 181415 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 697347 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 540060 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 89746 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P160513 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_65_P18756 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P271068 | BC064748.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P271068 | AK018361.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P271068 | AK139827.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P271068 | NM_029416.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P271068 | AF508984.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P271068 | BC064748.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P271068 | AK018361.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P271068 | AK139827.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P271068 | NM_029416.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P271068 | AF508984.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P271068 | BC064748.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P271068 | AK018361.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P271068 | AK139827.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P271068 | NM_029416.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P271068 | AF508984.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P271068 | BC064748.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P271068 | AK018361.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P271068 | AK139827.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P271068 | NM_029416.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P271068 | AF508984.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23236369 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23236384 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23236382 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23236383 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23236370 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23236394 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23236359 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23236387 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23236362 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23236363 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23236376 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23236367 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23236392 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23236389 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23236371 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23236379 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23236391 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23236374 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23236378 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23236364 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P160513 | - | - |
| Whole Mouse Genome G4122A | A_51_P271068 | BC064748.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P271068 | AK018361.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P271068 | AK139827.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P271068 | NM_029416.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P271068 | AF508984.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P271068 | BC064748.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P271068 | AK018361.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P271068 | AK139827.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P271068 | NM_029416.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P271068 | AF508984.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P160513 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03244088 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03435202 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03858358 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03858360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23236316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23236351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23236369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23236350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23236341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23236348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23236370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23236380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23236402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23236356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23236357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23236383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23236379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23236374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23236392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23236354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23236382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23236403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23236387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23236358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23236395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23236337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23236398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23236406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23236371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23236362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23236389 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23236376 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03943396 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23236398 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23236354 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23236379 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03834270 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23236337 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23236358 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23236387 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23236360 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23236340 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23236329 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23236406 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23236364 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03943395 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23236361 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23236378 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23236374 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23236389 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23236402 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23236385 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23236352 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23236341 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23236382 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04045238 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03329334 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03244087 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23236403 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23236380 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23236353 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23236384 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03599267 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23236363 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23236393 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23236392 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23236357 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23236356 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23236371 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23236391 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23236370 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23236348 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23236343 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23236394 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23236362 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23236359 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23236369 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10515496 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515497 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515499 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515498 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1456561_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438899_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429303_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10515495 | - | - |
| Mouse Genome 430 2.0 Array | 1429303_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1429303_at | - | - |
| Mouse Genome 430 2.0 Array | 1456561_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1456561_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1438899_at | - | - |
| Mouse Genome 430 2.0 Array | 1438899_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75753_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75753_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400027091.mm.1 | TC0400003375.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400027091.mm.1 | NM_029416.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400027091.mm.1 | NM_029416.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400027093.mm.1 | TC0400003375.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400027093.mm.1 | NM_029416.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400027093.mm.1 | NM_029416.2 | Refseq |
| Mouse Expression Set 430 | 1429303_PM_at | - | - |
| Mouse Expression Set 430 | 1429303_at | - | - |
| Murine Genome U74v2 Set | 162265_r_at | - | - |
| Murine Genome U74v2 Set | 92324_at | - | - |
| Murine Genome U74v2 Set | 92324_PM_at | - | - |
| Murine Genome U74v2 Set | 162265_PM_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1429303_at | - | - |
| Mouse Genome 430A 2.0 Array | 1429303_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1429303_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1429303_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456561_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438899_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438899_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1456561_PM_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 432414 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2320154 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3830095 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2320154 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6590156 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006590156 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003830095 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002320154 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2469125 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1227929 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1226178 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1227929 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2469125 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1226178 | - | - |
| mouse-6 v1.1 (Illumina) | scl0075753.2_179-S | - | - |
| mouse-6 v1.1 (Illumina) | scl075753.2_67-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0075753.2_78-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3830095 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6590156 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2320154 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0075753.2_78-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0075753.2_179-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl075753.2_67-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2320154 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6590156 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3830095 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1226178 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2469125 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1225165 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1225165 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2469125 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1226178 | - | - |

Klf17
Genomic Map