Assay Details
| Gene Symbol: Klhl24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029436.3 | NP_083712.4 | 1 - 2 | 120 | 134 | |
| GenBank mRNA | AK015239.1 | - | 1 - 2 | 106 | 134 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK015239.1,NM_029436.3 transcripts.
Set Membership: |
|

Klhl24
Genomic Map