Assay Details
| Gene Symbol: Lmf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029624.4 | NP_083900.3 | 7 - 8 | 1097 | 93 | |
| NR_045520.1 | 5 - 6 | 776 | 93 | |||
| GenBank mRNA | AK010312.1 | - | 7 - 8 | 1068 | 93 | |
| AK041659.1 | - | 5 - 6 | 764 | 93 | ||
| AK154622.1 | - | 7 - 8 | 1094 | 93 | ||
| AK169983.1 | - | 7 - 8 | 1095 | 93 | ||
| AK177501.1 | - | 2 - 3 | 230 | 94 | ||
| BC020104.1 | - | 7 - 8 | 1104 | 3963699 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lmf1
Genomic Map