Assay Details
| Gene Symbol: Glt8d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001165930.1 | NP_001159402.1 | 5 - 6 | 522 | 138 | |
| NM_029626.2 | NP_083902.2 | 5 - 6 | 1051 | 138 | ||
| GenBank mRNA | AK010388.1 | - | 5 - 6 | 780 | 138 | |
| AK077388.1 | - | 5 - 6 | 515 | 138 | ||
| AK160461.1 | - | 6 - 7 | 682 | 138 | ||
| BC055709.1 | - | 5 - 6 | 808 | 138 | ||
| BC058731.1 | - | 5 - 6 | 808 | 138 | ||
| BC069873.1 | - | 5 - 6 | 601 | 6314675 | 138 | |
| CX209391.1 | - | 4 - 5 | 482 | 138 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Glt8d1
Genomic Map