Assay Details
Gene Symbol: Spatc1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029661.1 | NP_083937.1 | 1 - 2 | 294 | 91 | |
GenBank mRNA | AK006410.1 | - | 1 - 2 | 294 | 91 | |
BC049686.1 | - | 1 - 2 | 285 | 91 | ||
BC147585.1 | - | 1 - 2 | 274 | 9056575 | 91 | |
BC147614.1 | - | 1 - 2 | 274 | 9056604 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2047827 | BC147614.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2047827 | BC147585.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2047827 | NM_029661.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2047827 | AK006410.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2047827 | BC049686.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P474288 | BC147614.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P474288 | BC147585.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P474288 | NM_029661.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P474288 | AK006410.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P474288 | BC049686.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P474288 | BC147614.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P474288 | BC147585.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P474288 | NM_029661.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P474288 | AK006410.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P474288 | BC049686.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 433784 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 42054 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 825854 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 125592 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 174028 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 145564 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P121604 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2047827 | BC147614.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2047827 | BC147585.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2047827 | NM_029661.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2047827 | AK006410.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2047827 | BC049686.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P474288 | BC147614.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P474288 | BC147585.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P474288 | NM_029661.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P474288 | AK006410.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P474288 | BC049686.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P474288 | BC147614.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P474288 | BC147585.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P474288 | NM_029661.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P474288 | AK006410.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P474288 | BC049686.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27262731 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27262766 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27262742 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27262759 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27262719 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27262763 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27262748 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27262762 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27262752 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27262746 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27262727 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27262744 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27262734 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27262738 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27262723 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27262755 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27262740 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27262725 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27262729 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27262735 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27262768 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27262722 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27262765 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27262757 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27262732 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27262760 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27262767 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27262736 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27262749 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27262721 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P121604 | - | - |
Whole Mouse Genome G4122A | A_51_P474288 | BC147614.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P474288 | BC147585.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P474288 | NM_029661.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P474288 | AK006410.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P474288 | BC049686.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P474288 | BC147614.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P474288 | BC147585.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P474288 | NM_029661.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P474288 | AK006410.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P474288 | BC049686.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126592 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P121604 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08218159 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08218172 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08218164 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08218158 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08218165 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262778 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262738 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262785 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262795 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262732 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262847 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262719 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262794 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262766 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262734 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262782 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262763 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262803 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262755 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262839 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262816 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262801 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262772 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262781 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262705 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262837 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262835 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262822 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262843 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262784 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262740 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262831 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262829 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262699 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262721 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262698 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262767 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262742 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262797 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262746 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262727 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262757 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262856 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262708 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262774 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262709 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262848 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262702 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262752 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262789 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262765 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262806 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262775 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262703 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262759 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262715 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262799 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262812 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262749 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262809 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262855 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262786 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262707 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262744 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262791 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262818 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262828 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262820 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262857 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262800 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27262713 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27262774 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27262782 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08729421 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27262770 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27262722 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27262765 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08855542 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27262795 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27262711 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27262709 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27262797 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27262732 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27262755 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27262792 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08218160 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27262786 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27262705 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08413799 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08439495 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27262775 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27262778 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27262734 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08407150 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27262713 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08236979 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27262718 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27262763 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27262719 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27262772 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08236978 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27262721 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27262794 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27262749 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27262746 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27262752 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27262742 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27262791 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08729423 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27262717 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27262727 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27262785 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27262735 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08729420 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27262759 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27262748 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27262740 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27262707 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27262736 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08359731 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27262781 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27262784 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08223464 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27262789 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27262744 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27262715 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10364221 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10364220 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10364219 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10364218 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1432333_PM_a_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1456866_PM_x_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10364217 | - | - |
Mouse Genome 430 2.0 Array | 1432333_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1456866_x_at | - | - |
Mouse Genome 430 2.0 Array | 1456866_PM_x_at | - | - |
Mouse Genome 430 2.0 Array | 1432333_a_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 76573_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 76573_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1000005376.mm.1 | NM_029661.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000005376.mm.1 | NM_029661.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000005376.mm.1 | TC1000000745.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000005377.mm.1 | NM_029661.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000005377.mm.1 | NM_029661.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000005377.mm.1 | TC1000000745.mm.1 | MTA TC |
Mouse Expression Set 430 | 1456866_x_at | - | - |
Mouse Expression Set 430 | 1456866_PM_x_at | - | - |
Mouse Expression Set 430 | 1432333_PM_a_at | - | - |
Mouse Expression Set 430 | 1432333_a_at | - | - |
Murine Genome U74v2 Set | 165846_r_at | - | - |
Murine Genome U74v2 Set | 165846_PM_r_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1456866_PM_x_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1456866_x_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1432333_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1432333_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 634940 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 940537 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000940537 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1255463 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1255463 | - | - |
mouse-6 v1.1 (Illumina) | scl38744.4.1_76-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 940537 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl38744.4.1_76-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 940537 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1255463 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1255463 | - | - |
