Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000005380.mm.1 |
NM_029661.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005380.mm.1 |
NM_029661.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005380.mm.1 |
TC1000000745.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000005381.mm.1 |
NM_029661.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005381.mm.1 |
NM_029661.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005381.mm.1 |
TC1000000745.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000005378.mm.1 |
NM_029661.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005378.mm.1 |
NM_029661.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005378.mm.1 |
TC1000000745.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000005382.mm.1 |
NM_029661.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005382.mm.1 |
NM_029661.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005382.mm.1 |
TC1000000745.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000005379.mm.1 |
NM_029661.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005379.mm.1 |
NM_029661.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005379.mm.1 |
TC1000000745.mm.1 |
MTA TC |