Assay Details
| Gene Symbol: Gsdma2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029727.2 | NP_084003.2 | 1 - 2 | 95 | 60 | |
| GenBank mRNA | AK008613.1 | - | 1 - 2 | 92 | 60 | |
| AK008663.1 | - | 1 - 2 | 92 | 60 | ||
| AK008685.1 | - | 1 - 2 | 92 | 60 | ||
| BC109336.1 | - | 1 - 2 | 29 | 40052146 | 60 | |
| BY090779.1 | - | 1 - 2 | 95 | 60 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK008613.1,AK008685.1,BC109336.1,NM_029727.2 transcripts.
Set Membership: |
|

Gsdma2
Genomic Map