Assay Details
| Gene Symbol: Mtfr1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256112.1 | NP_001243041.1 | 6 - 7 | 1064 | 88 | |
| NM_029759.4 | NP_084035.1 | 6 - 7 | 1015 | 88 | ||
| GenBank mRNA | AK010825.1 | - | 6 - 7 | 1056 | 88 | |
| AK145362.1 | - | 6 - 7 | 1056 | 88 | ||
| AK146416.1 | - | 5 - 6 | 891 | 88 | ||
| AK164004.1 | - | 5 - 6 | 891 | 88 | ||
| AK169047.1 | - | 6 - 7 | 923 | 88 | ||
| AK171703.1 | - | 6 - 7 | 1056 | 88 | ||
| BC026808.1 | - | 5 - 6 | 887 | 4159289 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23323336 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23323333 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23323340 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23323335 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23323341 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23323334 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23323339 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23323338 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1957098 | AK169047.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1957098 | AK089141.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1957098 | NM_029759.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2089780 | BC026808.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2089780 | AK169047.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2089780 | AK171703.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2089780 | AK187934.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2089780 | AK145362.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 558544 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 778613 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 125006 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 41811 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 288727 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 367047 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 34259 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P128392 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1957098 | AK169047.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1957098 | AK089141.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1957098 | NM_029759.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2089780 | BC026808.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2089780 | AK169047.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2089780 | AK171703.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2089780 | AK187934.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2089780 | AK145362.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23323318 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23323337 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23323330 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23323341 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23323344 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23323311 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23323327 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23323299 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23323336 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23323353 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23323313 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23323317 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23323297 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23323302 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23323323 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23323308 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23323343 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23323338 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23323309 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23323321 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23323314 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23323342 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23323347 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23323351 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23323304 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23323300 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23323306 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23323339 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23323333 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23323349 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23323325 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23323319 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05412177 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P128392 | - | - |
| Whole Mouse Genome G4122A | A_51_P453715 | BC026808.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P453715 | AK194688.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P453715 | AK196748.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P453715 | AK146416.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P33071 | BC026808.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P33071 | AK169047.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P33071 | AK171703.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P33071 | AK145362.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P33071 | AK146416.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120109 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P128392 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03263070 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03263069 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03263066 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03263084 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03263062 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03263081 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23323335 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23323341 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23323338 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23323334 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23323340 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23323333 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23323339 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23323336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23323270 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323347 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323302 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323275 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03263078 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323352 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323283 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323341 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323308 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323314 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323329 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323351 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323279 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323297 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323327 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323286 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323300 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323306 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323277 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03399650 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323333 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323274 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323284 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323289 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03669202 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323325 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03942486 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323319 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03399649 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03975522 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323304 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323266 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323282 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323313 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323323 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323343 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323311 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323342 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323349 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323269 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323321 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323292 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04028269 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323291 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323285 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23323354 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10517271 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517272 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517273 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517265 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517269 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517264 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517267 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517270 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517268 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517266 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420059_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426374_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420058_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10517263 | - | - |
| Mouse Genome 430 2.0 Array | 1420059_at | - | - |
| Mouse Genome 430 2.0 Array | 1420058_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1420059_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1426374_at | - | - |
| Mouse Genome 430 2.0 Array | 1420058_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1426374_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 76824_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 76824_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400030785.mm.1 | TC0400003740.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400030785.mm.1 | NM_001256112.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400030785.mm.1 | NM_001256112.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400030785.mm.1 | NM_029759.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400030785.mm.1 | NM_029759.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400030782.mm.1 | TC0400003740.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400030782.mm.1 | NM_001256112.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400030782.mm.1 | NM_001256112.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400030782.mm.1 | NM_029759.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400030782.mm.1 | NM_029759.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400030786.mm.1 | TC0400003740.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400030786.mm.1 | NM_001256112.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400030786.mm.1 | NM_001256112.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400030786.mm.1 | NM_029759.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400030786.mm.1 | NM_029759.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400030784.mm.1 | TC0400003740.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400030784.mm.1 | NM_001256112.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400030784.mm.1 | NM_001256112.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400030784.mm.1 | NM_029759.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400030784.mm.1 | NM_029759.4 | Refseq |
| Mouse Expression Set 430 | 1426374_PM_at | - | - |
| Mouse Expression Set 430 | 1426374_at | - | - |
| Murine Genome U74v2 Set | 96917_PM_at | - | - |
| Murine Genome U74v2 Set | 96917_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426374_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426374_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426374_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420059_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420059_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426374_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420058_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420058_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2650494 | - | - |
| MouseWG-6 v2 Expression BeadChip | 3840427 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2650494 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3840427 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002650494 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003840427 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2629717 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1216349 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2629717 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1216349 | - | - |
| mouse-6 v1.1 (Illumina) | scl0076824.1_187-S | - | - |
| mouse-6 v1.1 (Illumina) | scl23686.4_28-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2650494 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3840427 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl23686.4_28-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0076824.1_187-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3840427 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2650494 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2629718 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2629718 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0076824.1_187-S | - | - |

Mtfr1l
Genomic Map