Assay Details
| Gene Symbol: Hnrnpm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109913.1 | NP_001103383.1 | 12 - 13 | 1000 | 65 | |
| NM_029804.3 | NP_084080.1 | 11 - 12 | 1117 | 65 | ||
| GenBank mRNA | AK011521.1 | - | 11 - 12 | 1063 | 65 | |
| AK133911.1 | - | 11 - 12 | 1052 | 65 | ||
| AK145343.1 | - | 12 - 13 | 938 | 65 | ||
| AK159848.1 | - | 12 - 13 | 932 | 65 | ||
| AK166670.1 | - | 11 - 12 | 1052 | 65 | ||
| AK168347.1 | - | 12 - 13 | 935 | 65 | ||
| AK169101.1 | - | 12 - 13 | 935 | 65 | ||
| AK171892.1 | - | 12 - 13 | 935 | 65 | ||
| AK220299.1 | - | 12 - 13 | 960 | 65 | ||
| BC017539.1 | - | 12 - 13 | 930 | 65 | ||
| BC035525.1 | - | 12 - 13 | 935 | 65 | ||
| BC054785.1 | - | 2519 | 65 | |||
| BC065172.1 | - | 12 - 13 | 930 | 6415170 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hnrnpm
Genomic Map